Learn More
Medchemexpress LLC N-(3,5-dichloro-2-methoxyphenyl)-4-methoxy-3-(1-piperazinyl)-benzenesulfonamide monohydrochloride | 402713-81-9 | >98.0% | 482.8 g·mol⁻¹ | C18H21Cl2N3O4S · HCl | 25 MG
SDP
Catalog No. 5000246736
Encompass_Preferred
SB-399885 hydrochloride is a research compound that acts as a selective antagonist of the serotonin 5-HT6 receptor (CAS 402713-81-9). It is used in vitro and in vivo to study 5-HT6-mediated signaling and cognitive effects, and is supplied as the monohydrochloride salt to improve solubility for biological assays.
  • 5-HT6 receptor antagonist activity for receptor pharmacology studies.
  • Reported high purity for reliable experimental results.
  • Monohydrochloride salt form improves solubility in aqueous media.
  • Suitable for in vitro and in vivo neuroscience applications.
  • Supplied in small research pack sizes for dose-response and pilot studies.
Catalog No. 50-002-46736 Supplier Medchemexpress LLC Supplier No. HY10309925MG
SB-399885 hydrochloride is a research compound that acts as a selective antagonist of the serotonin 5-HT6 receptor (CAS 402713-81-9). It is used in vitro and in vivo to study 5-HT6-mediated signaling and cognitive effects, and is supplied as the monohydrochloride salt to improve solubility for biological assays.
  • 5-HT6 receptor antagonist activity for receptor pharmacology studies.
  • Reported high purity for reliable experimental results.
  • Monohydrochloride salt form improves solubility in aqueous media.
  • Suitable for in vitro and in vivo neuroscience applications.
  • Supplied in small research pack sizes for dose-response and pilot studies.