Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(3,5-dimethylphenyl)-N'-[2-(trifluoromethyl)phenyl]-guanidine | 1430208-73-3 | MFCD31619254 | 99.7% | 307.31 | C16H16F3N3 | 1 ML
SDP

Catalog No. 5000242649
Encompass_Preferred

1A-116 is a small-molecule Rac1 inhibitor used in biological research to block Rac1 activation and downstream signaling. It disrupts Rac1-P-Rex1 interaction, induces apoptosis, and inhibits cell proliferation, migration, and cell cycle progression, and has shown antimetastatic activity in vivo.

  • Potent Rac1 inhibitor that blocks Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation, migration, and cell cycle progression.
  • Demonstrated antimetastatic activity in vivo.
  • Available as a 10 mM solution in DMSO (1 mL) and in solid formats.
  • High purity, 99.7%.
  • Molecular weight 307.31 g/mol and formula C16H16F3N3.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Catalog No. 50-002-42649 Supplier Medchemexpress LLC Supplier No. HY10406410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1A-116 is a small-molecule Rac1 inhibitor used in biological research to block Rac1 activation and downstream signaling. It disrupts Rac1-P-Rex1 interaction, induces apoptosis, and inhibits cell proliferation, migration, and cell cycle progression, and has shown antimetastatic activity in vivo.

  • Potent Rac1 inhibitor that blocks Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation, migration, and cell cycle progression.
  • Demonstrated antimetastatic activity in vivo.
  • Available as a 10 mM solution in DMSO (1 mL) and in solid formats.
  • High purity, 99.7%.
  • Molecular weight 307.31 g/mol and formula C16H16F3N3.
  • Recommended storage: powder at -20°C; in solvent at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.