Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(-)-N-(3,5-Dinitrobenzoyl)-Alpha-Phenylglycine

Catalog No. 503081431 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-308-1431 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-308-1431 Supplier Sigma Aldrich Supplier No. 2500315G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-(-)-N-(3,5-Dinitrobenzoyl)-Alpha-Phenylglycine

Specifications

CAS 13170-43-9
Density 0.777 g/mL at 25°C
Molecular Formula C4H11ClMgSi
Linear Formula (CH3)3SiCH2MgCl
MDL Number MFCD00009916
Quantity 100 mL
Molecular Weight (g/mol) 146.97 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.