Learn More
Medchemexpress LLC N-[3-[[6-fluoro-2-[4-(4-methylpiperazin-1-yl)anilino]quinazolin-4-yl]amino]phenyl]prop-2-enamide | 2955607-40-4 | 99.7% | 497.57 g·mol⁻1 | C28H28FN7O | 10 MG
SDP
Catalog No. 5000285793
Encompass_Preferred
A small-molecule inhibitor selective for the epidermal growth factor receptor (EGFR) double mutants T790M and L858R, supplied as a research reagent for in vitro biochemical and cellular studies. It exhibits mutant-selective inhibition and is provided at high chemical purity for assay work.
  • Intended for research use only; not for human or veterinary use.
  • Selective inhibitor of EGFR T790M and L858R mutants.
  • Reported IC50 ≈ 3.5 nM for mutant EGFR and ≈ 1290 nM for wild-type EGFR.
  • High chemical purity (~99.7%).
  • Molecular formula C28H28FN7O and molecular weight 497.57 g·mol⁻1.
  • Available in milligram pack sizes suitable for biochemical and cellular assays.
Catalog No. 50-002-85793 Supplier Medchemexpress LLC Supplier No. HY14982410MG
A small-molecule inhibitor selective for the epidermal growth factor receptor (EGFR) double mutants T790M and L858R, supplied as a research reagent for in vitro biochemical and cellular studies. It exhibits mutant-selective inhibition and is provided at high chemical purity for assay work.
  • Intended for research use only; not for human or veterinary use.
  • Selective inhibitor of EGFR T790M and L858R mutants.
  • Reported IC50 ≈ 3.5 nM for mutant EGFR and ≈ 1290 nM for wild-type EGFR.
  • High chemical purity (~99.7%).
  • Molecular formula C28H28FN7O and molecular weight 497.57 g·mol⁻1.
  • Available in milligram pack sizes suitable for biochemical and cellular assays.