Learn More
Strem, An Ascensus Company CAS# 851051-43-9. 500mg. {N-[3-(η;6-phenyl)propyl]-[(1S-2S)-1,2-diphenyl-1-4-methylbenzenesulfonylamidato(kN’)-ethyl-2-amino-(kN)]}ruthenium(II) (S,S)-Teth-TsDpen RuCl WILLS CATALYST. MFCD16294984
SDP
Supplier:  Strem, An Ascensus Company 440110500MG
Encompass
CAS# 851051-43-9. 500mg. {N-[3-(η;6-phenyl)propyl]-[(1S-2S)-1,2-diphenyl-1-4-methylbenzenesulfonylamidato(kN’)-ethyl-2-amino-(kN)]}ruthenium(II) (S,S)-Teth-TsDpen RuCl WILLS CATALYST. MFCD16294984. Molecular Weight: 620.17. Molecular Formula: C30H31ClN2O2RuS. Color/form: orange pwdr. Strem# 44-0110. http://www.strem.com/catalog/v/44-0110/
Catalog No. 50-372-2