Learn More
Medchemexpress LLC N-(3-Acetyl-4-(2-hydroxy-3-(isopropylamino)propoxy-1,1,2,3,3-d5)phenyl)butyramide | 1189500-68-2 | 897231 | 99.7% | 341.46 | 1 MG
SDP
Catalog No. 5000387002
Encompass_Preferred
Acebutolol-d5 is the deuterium labeled Acebutolol. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87002 Supplier Medchemexpress LLC Supplier No. HY17497S11MG
Acebutolol-d5 is the deuterium labeled Acebutolol. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.