Learn More
Medchemexpress LLC N'-(3-acetylphenyl)-N-(3-(2-methoxyethyl)-2,4-dioxo-1-(2-(piperidin-1-yl)ethyl)-1,2,3,4-tetrahydroquinazolin | 2946701-09-1 | 99.3% | 507.58 g/mol | C27H33N5O5 | 10 MG
SDP
Catalog No. 5000288329
Encompass_Preferred
COP1-ATGL modulator 1 is an orally active small-molecule research reagent that modulates the COP1-ATGL pathway. It increases ATGL protein expression, reduces ATGL and COP1 ubiquitination, and diminishes lipid accumulation in hepatocytes at nanomolar concentrations. Supplied as a white to off-white solid with high purity and defined storage recommendations for powder and solution.
  • Orally active modulator of COP1-ATGL signaling.
  • Increases ATGL protein expression at nanomolar concentrations.
  • Reduces ATGL and COP1 ubiquitination in cellular assays.
  • High purity suitable for research applications.
  • Stable solid with recommended low-temperature storage.
Catalog No. 50-002-88329 Supplier Medchemexpress LLC Supplier No. HY15741610MG
COP1-ATGL modulator 1 is an orally active small-molecule research reagent that modulates the COP1-ATGL pathway. It increases ATGL protein expression, reduces ATGL and COP1 ubiquitination, and diminishes lipid accumulation in hepatocytes at nanomolar concentrations. Supplied as a white to off-white solid with high purity and defined storage recommendations for powder and solution.
  • Orally active modulator of COP1-ATGL signaling.
  • Increases ATGL protein expression at nanomolar concentrations.
  • Reduces ATGL and COP1 ubiquitination in cellular assays.
  • High purity suitable for research applications.
  • Stable solid with recommended low-temperature storage.