Learn More
Medchemexpress LLC N-(3-(aminomethyl)benzyl)-8-(4-benzylpiperazin-1-yl)pyrimido[5,4-d]pyrimidin-4-amine | 1875036-75-1 | >98.0% | 25 MG
SDP
Catalog No. 5000232153
Encompass_Preferred
BI-8626 is a small-molecule inhibitor of the HECT E3 ubiquitin ligase HUWE1, identified by high-throughput screening and reported to inhibit HUWE1 with an IC50 of approximately 0.9 μM. It is supplied for research use only and intended for biochemical and cellular studies.
  • Specific HUWE1 inhibitor, IC50 ≈ 0.9 μM.
  • Used in biochemical and cellular assays to modulate HUWE1-dependent ubiquitination.
  • CAS 1875036-75-1; molecular weight 440.54 g·mol⁻¹.
  • Typical reported purity ≥98% (HPLC); verify certificate of analysis per lot.
  • Available as solid and as 10 mM solution in DMSO for reconstitution.
Catalog No. 50-002-32153 Supplier Medchemexpress LLC Supplier No. HY12020425MG
BI-8626 is a small-molecule inhibitor of the HECT E3 ubiquitin ligase HUWE1, identified by high-throughput screening and reported to inhibit HUWE1 with an IC50 of approximately 0.9 μM. It is supplied for research use only and intended for biochemical and cellular studies.
  • Specific HUWE1 inhibitor, IC50 ≈ 0.9 μM.
  • Used in biochemical and cellular assays to modulate HUWE1-dependent ubiquitination.
  • CAS 1875036-75-1; molecular weight 440.54 g·mol⁻¹.
  • Typical reported purity ≥98% (HPLC); verify certificate of analysis per lot.
  • Available as solid and as 10 mM solution in DMSO for reconstitution.