Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(3-aminophenyl)methyl]-6-[4-[(3-fluorophenyl)methoxy]phenoxy]pyridine-3-carboxamide | 838819-70-8 | MFCD28160863 | 99.2% | C26H22FN3O3 | 10MG
SDP

Catalog No. 5000260319
Encompass_Preferred

YM-244769 is a potent, selective, orally active Na+/Ca2+ exchanger (NCX) inhibitor with preferential activity against NCX3, used in research to investigate NCX function and calcium handling in cellular and in vivo studies.

  • Potent and selective NCX inhibitor with preference for NCX3.
  • High purity: 99.2%.
  • Solid, off-white to light yellow appearance.
  • Soluble in DMSO (≈120 mg/mL); suitable for in vitro use and for formulation in vivo studies.
  • Storage: sealed at 4°C; solutions stable at -80°C for up to 6 months.
  • Available in small research pack sizes to support experimental workflows.

Catalog No. 50-002-60319 Supplier Medchemexpress LLC Supplier No. HY136182A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

YM-244769 is a potent, selective, orally active Na+/Ca2+ exchanger (NCX) inhibitor with preferential activity against NCX3, used in research to investigate NCX function and calcium handling in cellular and in vivo studies.

  • Potent and selective NCX inhibitor with preference for NCX3.
  • High purity: 99.2%.
  • Solid, off-white to light yellow appearance.
  • Soluble in DMSO (≈120 mg/mL); suitable for in vitro use and for formulation in vivo studies.
  • Storage: sealed at 4°C; solutions stable at -80°C for up to 6 months.
  • Available in small research pack sizes to support experimental workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.