Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)-2-nitrobenzenesulfonamide Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A22681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22681G 1 g
A22685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22681G Supplier TCI America Supplier No. A22681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 863983-46-4
Molecular Formula C9H14ClN3O4S
Molecular Weight (g/mol) 295.74
MDL Number MFCD07366920
InChI Key UADJAJMVAIAKHQ-UHFFFAOYSA-N
Synonym 1-Amino-3-(2-nitrobenzenesulfonamido)propane Hydrochloride, N-(2-Nitrobenzenesulfonyl)-1,3-propanediamine Hydrochloride, N-(o-Ns)-1,3-diaminopropane Hydrochloride
PubChem CID 44630012
IUPAC Name N-(3-aminopropyl)-2-nitrobenzene-1-sulfonamide hydrochloride
SMILES Cl.NCCCNS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 295.74
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(3-Aminopropyl)-2-nitrobenzenesulfonamide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.