Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(3-Aminopropyl)-2-pyrrolidinone

Catalog No. 1365655G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-656-55G 5 g
13-656-525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-656-55G Supplier Sigma Aldrich Supplier No. 1365655G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-(3-aminopropyl)-2-pyrrolidinone (pyr) conjugates (vpa-pyr) of valproic acid (va) have been investigated for their anticonvulsant activity in mice
  • It is the α-lactam form of isoputreanine.n-(3-aminopropyl)-2-pyrrolidinone was used in the simultaneous determination of total polyamines and three of their nonα amino acid metabolites in urine by capillary gas-chromatographic method with nitrogen-phosphorus detection.contains varying amounts of 1,5-diazabicyclo[4.3.0]non-5-ene (cat
  • No; 136581)

Specifications

CAS 7663-77-6
Density 1.014 g/mL (at 25°C (literature))
Boiling Point 120°C to 123°C (1 mmHg)
Molecular Formula C7H14N2O
Refractive Index n20/D 1.5 (literature)
Linear Formula C7H14N2O
MDL Number MFCD00003201
Quantity 5 g
Molecular Weight (g/mol) 142.2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.