Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(3-aminopropyl)cyclohexylamine | 3312-60-5 | MFCD00003838 | 98.8% | 156.27 g/mol | C9H20N2 | 1 ML
SDP

Catalog No. 5000302959
Encompass_Preferred

N-(3-Aminopropyl)cyclohexylamine (CAS 3312-60-5) is a small-molecule spermidine synthase inhibitor supplied as a liquid or ready-to-use solution for biochemical and pharmacological research. It is used in assays probing polyamine metabolism and enzyme inhibition, and is offered in solution formats for direct use in experiments.

  • High purity (98.8%).
  • Molecular weight 156.27 g/mol.
  • Chemical formula C9H20N2.
  • Appearance: colorless to light yellow liquid.
  • Soluble in DMSO, water, and PBS; may require ultrasonic treatment.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-02959 Supplier Medchemexpress LLC Supplier No. HYW01551410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-(3-Aminopropyl)cyclohexylamine (CAS 3312-60-5) is a small-molecule spermidine synthase inhibitor supplied as a liquid or ready-to-use solution for biochemical and pharmacological research. It is used in assays probing polyamine metabolism and enzyme inhibition, and is offered in solution formats for direct use in experiments.

  • High purity (98.8%).
  • Molecular weight 156.27 g/mol.
  • Chemical formula C9H20N2.
  • Appearance: colorless to light yellow liquid.
  • Soluble in DMSO, water, and PBS; may require ultrasonic treatment.
  • Storage: 4°C protected from light; in solvent store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.