Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Aminopropyl)cyclohexylamine 98.0+%, TCI America™
SDP

Catalog No. A0891500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0891500ML 500 mL
A089125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0891500ML Supplier TCI America Supplier No. A0891500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3312-60-5
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.273
MDL Number MFCD00003838
InChI Key ITZPOSYADVYECJ-UHFFFAOYSA-N
Synonym n-3-aminopropyl cyclohexylamine, n1-cyclohexylpropane-1,3-diamine, 1,3-propanediamine, n-cyclohexyl, n-cyclohexyl-1,3-propanediamine, cyclohexyl-1,3-propanediamine, 3-aminopropyl cyclohexylamine, n-cyclohexyl-1,3-diaminopropane, unii-f4xob09fkn, n-cyclohexyl-1,3-propylenediamine, n-cyclohexylpropylene 1,3-diamine
PubChem CID 18718
IUPAC Name N'-cyclohexylpropane-1,3-diamine
SMILES C1CCC(CC1)NCCCN

Specifications

Color Yellow
Boiling Point 123°C
Quantity 500 mL
UN Number 2735
Formula Weight 156.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(3-Aminopropyl)cyclohexylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.