Learn More
Medchemexpress LLC N-[(3-(anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride | 63857-00-1 | 96.5% | 359.29 | C20H20Cl2N2 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01930650G
Encompass_Preferred
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an organic intermediate used as a building block in the synthesis of cyanine-containing dyes and related compounds. Supplied as a white crystalline solid for research use only, it has a documented molecular formula and molecular weight, and safety data sheets are provided by the manufacturer.
Catalog No. 50-003-00916