Learn More
Medchemexpress LLC N-[(3-(anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride | 63857-00-1 | 96.5% | 359.29 | C20H20Cl2N2 | 500 G
SDP
Catalog No. 5000299945
Encompass_Preferred
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an organic intermediate supplied as the monohydrochloride salt for research and analytical synthesis. It is used primarily as a building block in the preparation of cyanine-containing dyes and other conjugated chromophores and is offered in laboratory and bulk pack sizes.
  • Monohydrochloride salt form for handling and stability.
  • High purity (96.5% assay).
  • Molecular weight 359.29 g/mol; formula C20H20Cl2N2.
  • Suitable for synthesis of cyanine dyes and related conjugated compounds.
  • Available in multiple pack sizes, including larger quantities by quotation.
  • Supplied with a product datasheet containing technical and safety information.
Catalog No. 50-002-99945 Supplier Medchemexpress LLC Supplier No. HYW019306500G
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an organic intermediate supplied as the monohydrochloride salt for research and analytical synthesis. It is used primarily as a building block in the preparation of cyanine-containing dyes and other conjugated chromophores and is offered in laboratory and bulk pack sizes.
  • Monohydrochloride salt form for handling and stability.
  • High purity (96.5% assay).
  • Molecular weight 359.29 g/mol; formula C20H20Cl2N2.
  • Suitable for synthesis of cyanine dyes and related conjugated compounds.
  • Available in multiple pack sizes, including larger quantities by quotation.
  • Supplied with a product datasheet containing technical and safety information.