Learn More
Medchemexpress LLC N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride | 63857-00-1 | 96.5% | 359.29 g·mol-1 | C20H20Cl2N2 | 10 G
SDP
Catalog No. 5000299944
Encompass_Preferred
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an aniline-derived chemical building block provided as the hydrochloride salt for research and synthetic applications. It is used primarily to synthesize cyanine-containing and related conjugated dyes and is supplied as a solid with a reported purity of 96.5%.
  • Aniline-derived building block for dye synthesis.
  • Useful for synthesis of cyanine-containing and conjugated dyes.
  • Supplied as the hydrochloride salt, improving handling and stability.
  • Solid form, dark purple to black appearance.
  • High purity (96.5%), suitable for research and synthesis.
  • Molecular weight 359.29 g·mol-1; formula C20H20Cl2N2.
  • Available in multiple pack sizes to support small-scale and larger-scale work.
Catalog No. 50-002-99944 Supplier Medchemexpress LLC Supplier No. HYW01930610G
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an aniline-derived chemical building block provided as the hydrochloride salt for research and synthetic applications. It is used primarily to synthesize cyanine-containing and related conjugated dyes and is supplied as a solid with a reported purity of 96.5%.
  • Aniline-derived building block for dye synthesis.
  • Useful for synthesis of cyanine-containing and conjugated dyes.
  • Supplied as the hydrochloride salt, improving handling and stability.
  • Solid form, dark purple to black appearance.
  • High purity (96.5%), suitable for research and synthesis.
  • Molecular weight 359.29 g·mol-1; formula C20H20Cl2N2.
  • Available in multiple pack sizes to support small-scale and larger-scale work.