Learn More
Cayman Chemical (±)-N-3-Benzylnirvanol, Purity: ≥98%, CAS: 93879-40-4, 10 mg
SDP
Catalog No. 502940898
Encompass_Preferred
A racemic mixture of (+)-N-3-benzylnirvanol and (–)-N-3-benzylnirvanol; lowers plasma levels of cholesterol in mice
Catalog No. 50-294-0898 Supplier Cayman Chemical Supplier No. 2729810 MG
A racemic mixture of (+)-N-3-benzylnirvanol and (–)-N-3-benzylnirvanol; lowers plasma levels of cholesterol in mice