Learn More
Cayman Chemical (+)-N-3-Benzylnirvanol, Purity: ≥95%, CAS: 790676-40-3, 5 mg
SDP
Catalog No. 502989859
Encompass_Preferred
An inhibitor of CYP2C19 (Ki = 0.25 µM); selective for CYP2C19 over CYP1A2, -2A6, -2C8, -2C9, -2D6, -2E1, and -3A4 at 1 µM and over CYP2B6 (IC50 = 58 µM)
Catalog No. 50-298-9859 Supplier Cayman Chemical Supplier No. 300975 MG
An inhibitor of CYP2C19 (Ki = 0.25 µM); selective for CYP2C19 over CYP1A2, -2A6, -2C8, -2C9, -2D6, -2E1, and -3A4 at 1 µM and over CYP2B6 (IC50 = 58 µM)