Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(3-Bromophenyl)-3-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Catalog No. AAH57501MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH57501MD 250 mg
AAH5750103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH57501MD Supplier Thermo Scientific Chemicals Supplier No. H57501MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The derivatives of this compound were identified as highly potent Bcr-Abl kinase inhibitors that is used to treat chronic myeloid leukemia.

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials. Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 710310-54-6
Molecular Formula C14H9BrF3NO
Molecular Weight (g/mol) 344.131
InChI Key NUCGNVOUNFOXOQ-UHFFFAOYSA-N
Synonym n-3-bromophenyl-3-trifluoromethyl benzamide, n-3-bromophenyl 3-trifluoromethyl phenyl carboxamide
PubChem CID 2079846
IUPAC Name N-(3-bromophenyl)-3-(trifluoromethyl)benzamide
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)NC2=CC(=CC=C2)Br

Specifications

Quantity 250 mg
Formula Weight 344.13
Percent Purity 97%
Chemical Name or Material N-(3-Bromophenyl)-3-(trifluoromethyl)benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.