Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(3-bromophenyl)-4-hydroxy-2-oxo-1H-quinoline-3-carboxamide | 333419-97-9 | 99.2% | 359.17 g/mol | C16H11BrN2O3 | 10 MG
SDP

Catalog No. 5000289877
Encompass_Preferred

EGFR T790M/L858R-IN-8 is a small-molecule inhibitor selective for the EGFR T790M/L858R mutant, supplied as a solid for in vitro research use only. The compound has defined physicochemical properties, demonstrated activity against the mutant EGFR in biochemical assays, and is provided in milligram quantities for laboratory studies.

  • Selective EGFR T790M/L858R inhibition (IC50 56.8 μM).
  • Intended for research use only, not for human or clinical use.
  • Solid powder form; store powder at -20°C.
  • High purity: 99.2%.
  • Soluble in DMSO: 2 mg/mL (5.57 mM) with sonication and warming.
  • CAS number 333419-97-9; molecular formula C16H11BrN2O3; molecular weight 359.17 g/mol.
  • Available in milligram quantities suitable for laboratory experiments.

Catalog No. 50-002-89877 Supplier Medchemexpress LLC Supplier No. HY16405610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGFR T790M/L858R-IN-8 is a small-molecule inhibitor selective for the EGFR T790M/L858R mutant, supplied as a solid for in vitro research use only. The compound has defined physicochemical properties, demonstrated activity against the mutant EGFR in biochemical assays, and is provided in milligram quantities for laboratory studies.

  • Selective EGFR T790M/L858R inhibition (IC50 56.8 μM).
  • Intended for research use only, not for human or clinical use.
  • Solid powder form; store powder at -20°C.
  • High purity: 99.2%.
  • Soluble in DMSO: 2 mg/mL (5.57 mM) with sonication and warming.
  • CAS number 333419-97-9; molecular formula C16H11BrN2O3; molecular weight 359.17 g/mol.
  • Available in milligram quantities suitable for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.