Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[3-chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]-methanesulfon... | 1621175-65-2 | MFCD30182283 | 99.4% | 418.90 g/mol | C18H19ClN6O2S | 1 ML
SDP

Catalog No. 5000246933
Encompass_Preferred

TC-G-1008 (GPR39-C3) is a potent, orally available agonist of the GPR39 zinc-sensing G protein-coupled receptor, exhibiting EC50 values of 0.4 nM (rat) and 0.8 nM (human) and commonly used in in vitro and in vivo pharmacology studies.

  • Potent GPR39 agonist (EC50 0.4 nM rat, 0.8 nM human).
  • Orally available and active in in vivo models.
  • Available as 10 mM solution in DMSO (1 mL) or solid quantities.
  • Molecular formula C18H19ClN6O2S; molecular weight 418.90 g/mol.
  • Reported purity 99.4% (HPLC).
  • Intended for research use in pharmacology and metabolic studies.

Catalog No. 50-002-46933 Supplier Medchemexpress LLC Supplier No. HY10300710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TC-G-1008 (GPR39-C3) is a potent, orally available agonist of the GPR39 zinc-sensing G protein-coupled receptor, exhibiting EC50 values of 0.4 nM (rat) and 0.8 nM (human) and commonly used in in vitro and in vivo pharmacology studies.

  • Potent GPR39 agonist (EC50 0.4 nM rat, 0.8 nM human).
  • Orally available and active in in vivo models.
  • Available as 10 mM solution in DMSO (1 mL) or solid quantities.
  • Molecular formula C18H19ClN6O2S; molecular weight 418.90 g/mol.
  • Reported purity 99.4% (HPLC).
  • Intended for research use in pharmacology and metabolic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.