Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(3-chloro-4-(6-methoxypyrazolo[1,5-a]pyridine-3-carboxamido)phenyl)-1-(1-(3-cyano-3-methylbutanoyl | 2380027-49-4 | MFCD32263443 | 98.6% | 671.12 | C34H32ClFN8O4 | 5 MG
SDP

Catalog No. 5000263899
Encompass_Preferred

CSK-IN-1 is a research-grade small-molecule inhibitor of CSK/Src family tyrosine kinases supplied as a white powder for laboratory use. It corresponds to CAS 2380027-49-4, has molecular formula C34H32ClFN8O4 and molecular weight 671.12 Da. Typical pack sizes include 1 mg, 5 mg, and 10 mg. Storage and purity are batch-dependent and follow standard research-reagent handling recommendations.

  • Inhibits Src and related protein tyrosine kinases.
  • Supplied as a stable white powder for research use.
  • Molecular weight 671.12 Da and formula C34H32ClFN8O4.
  • Available in small lab-scale pack sizes (1 mg, 5 mg, 10 mg).
  • Recommended storage: powder at -20°C; in solvent at -80°C.
  • Batch-specific purity approximately 98.6%.
  • Suitable for in vitro biochemical and cellular assays.

Catalog No. 50-002-63899 Supplier Medchemexpress LLC Supplier No. HY1302545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CSK-IN-1 is a research-grade small-molecule inhibitor of CSK/Src family tyrosine kinases supplied as a white powder for laboratory use. It corresponds to CAS 2380027-49-4, has molecular formula C34H32ClFN8O4 and molecular weight 671.12 Da. Typical pack sizes include 1 mg, 5 mg, and 10 mg. Storage and purity are batch-dependent and follow standard research-reagent handling recommendations.

  • Inhibits Src and related protein tyrosine kinases.
  • Supplied as a stable white powder for research use.
  • Molecular weight 671.12 Da and formula C34H32ClFN8O4.
  • Available in small lab-scale pack sizes (1 mg, 5 mg, 10 mg).
  • Recommended storage: powder at -20°C; in solvent at -80°C.
  • Batch-specific purity approximately 98.6%.
  • Suitable for in vitro biochemical and cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.