Learn More
Sigma Aldrich Fine Chemicals Biosciences N 3 Dimethylaminopropyl N
Catalog No. 502907272
Encompass
N-(3-Dimethylaminopropyl)-Nethylcarbodiimide is commonly used in combination with N-hydroxysuccinimide (NHS) in carbodiimide coupling reaction to activate carboxyl group for coupling with amines to form amides.
Catalog No. 50-290-7272 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3939150ML
N-(3-Dimethylaminopropyl)-Nethylcarbodiimide is commonly used in combination with N-hydroxysuccinimide (NHS) in carbodiimide coupling reaction to activate carboxyl group for coupling with amines to form amides.