Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-(3-Dimethylaminopropyl)-N'-Ethylcarbodiimide

Catalog No. 3939110ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-391-10ML 10 mL
39-391-50ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 39-391-10ML Supplier Sigma Aldrich Supplier No. 3939110ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-(3-Dimethylaminopropyl)-N'sa-ethylcarbodiimide is commonly used in combination with N-hydroxysuccinimide (NHS) in carbodiimide coupling reaction to activate carboxyl group for coupling with amines to form amides.N-(3-Dimethylaminopropyl)-N'sa-ethylcarbodiimide has been used for the surface functionalization of graphene quantum dots (GQDs) employed as sensing probes in Janus micromotors to detect enterobacterial contamination. It has also been used for the activation of folic acid, prior to its conjugation on silica nanoparticles.Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. 39391.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Specifications

CAS 1892-57-5
Density 0.877 g/mL at 20°C
Molecular Formula C8H17N3
Refractive Index n20/D 1.461
Linear Formula C8H17N3
MDL Number MFCD00044916
Quantity 10 mL
Synonym N-Ethyl-N'-(3-dimethylaminopropyl)carbodiimide; EDC; WSC
Molecular Weight (g/mol) 155.24 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.