Learn More
Medchemexpress LLC N-(3-(imidazo[2,1-b]thiazol-6-yl)phenyl)-2-methoxybenzamide | 667910-69-2 | 99.2% | C19H15N3O2S | 10MG
SDP
Catalog No. 5000259537
Encompass_Preferred
Sirtuin modulator 2 (Compound 132) is a small-molecule research chemical that modulates sirtuin family activity. It is reported to have an ED50 ≤ 50 μM and is supplied as a high-purity solid intended for in vitro biochemical and cellular studies, target validation, and mechanism-of-action research.
  • Modulates sirtuin activity with ED50 ≤ 50 μM.
  • High purity suitable for research assays (99.2%).
  • Molecular formula C19H15N3O2S and molecular weight 349.41.
  • Supplied as a small-quantity solid for laboratory use (10 mg).
  • Suitable for biochemical and cellular assay applications.
Catalog No. 50-002-59537 Supplier Medchemexpress LLC Supplier No. HY14723410MG
Sirtuin modulator 2 (Compound 132) is a small-molecule research chemical that modulates sirtuin family activity. It is reported to have an ED50 ≤ 50 μM and is supplied as a high-purity solid intended for in vitro biochemical and cellular studies, target validation, and mechanism-of-action research.
  • Modulates sirtuin activity with ED50 ≤ 50 μM.
  • High purity suitable for research assays (99.2%).
  • Molecular formula C19H15N3O2S and molecular weight 349.41.
  • Supplied as a small-quantity solid for laboratory use (10 mg).
  • Suitable for biochemical and cellular assay applications.