Learn More
Medchemexpress LLC N-[3-[methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino]cyclobutyl]propane-1-sulfonamide | 1622902-68-4 | MFCD30187577 | 99.3% | 323.41 g·mol⁻¹ | C14H21N5O2S | 50 MG
SDP
Catalog No. 5000239757
Encompass_Preferred
Abrocitinib is a selective, orally active JAK1 inhibitor supplied as a research-grade reference standard for biochemical and cellular assays targeting the JAK/STAT pathway. It demonstrates potent inhibition of JAK1 and reduced activity against JAK2, and is intended for laboratory research use only (not for human or veterinary use).
  • Selective JAK1 inhibition with reported IC50 of 29 nM.
  • Reduced JAK2 activity (IC50 ≈ 803 nM).
  • High purity suitable for reference and assay use (purity 99.3%).
  • Molecular formula C14H21N5O2S; molecular weight 323.41 g·mol⁻¹.
  • Supplied for biochemical and cellular research applications.
  • Available in multiple package sizes, including 50 mg.
Catalog No. 50-002-39757 Supplier Medchemexpress LLC Supplier No. HY10742950MG
Abrocitinib is a selective, orally active JAK1 inhibitor supplied as a research-grade reference standard for biochemical and cellular assays targeting the JAK/STAT pathway. It demonstrates potent inhibition of JAK1 and reduced activity against JAK2, and is intended for laboratory research use only (not for human or veterinary use).
  • Selective JAK1 inhibition with reported IC50 of 29 nM.
  • Reduced JAK2 activity (IC50 ≈ 803 nM).
  • High purity suitable for reference and assay use (purity 99.3%).
  • Molecular formula C14H21N5O2S; molecular weight 323.41 g·mol⁻¹.
  • Supplied for biochemical and cellular research applications.
  • Available in multiple package sizes, including 50 mg.