Learn More
Medchemexpress LLC N-[3-oxo-3-[4-(4-pyridinyl)-1-piperazinyl]propyl]-2,1,3-benzothiadiazole-4-sulfonamide | 1135243-19-4 | MFCD18086889 | 99.8% | 432.52 g/mol | C18H20N6O3S2 | 25 MG
SDP
Catalog No. 5000244005
Encompass_Preferred
VU 0255035 is a selective, competitive antagonist of the M1 muscarinic acetylcholine receptor used as a research compound for central nervous system studies, including seizure models and investigations of neuronal excitability. It is supplied with high purity and is available in solid and solution formats for in vitro and in vivo experiments.
  • Selective M1 antagonist with reported nanomolar activity.
  • Brain-penetrant compound suitable for in vivo CNS studies.
  • High supplied purity (≈99.8%).
  • Molecular weight 432.52 g/mol (C18H20N6O3S2).
  • Available in multiple pack sizes and DMSO solution formats for easy dosing.
Catalog No. 50-002-44005 Supplier Medchemexpress LLC Supplier No. HY10823425MG
VU 0255035 is a selective, competitive antagonist of the M1 muscarinic acetylcholine receptor used as a research compound for central nervous system studies, including seizure models and investigations of neuronal excitability. It is supplied with high purity and is available in solid and solution formats for in vitro and in vivo experiments.
  • Selective M1 antagonist with reported nanomolar activity.
  • Brain-penetrant compound suitable for in vivo CNS studies.
  • High supplied purity (≈99.8%).
  • Molecular weight 432.52 g/mol (C18H20N6O3S2).
  • Available in multiple pack sizes and DMSO solution formats for easy dosing.