Learn More
Medchemexpress LLC N-[3-oxo-3-(4-pyridin-4-ylpiperazin-1-yl)propyl]-2,1,3-benzothiadiazole-4-sulfonamide | 1135243-19-4 | MFCD18086889 | 99.8% | 432.52 | C18H20N6O3S2 | 10 MG
SDP
Catalog No. 5000242004
Encompass_Preferred
VU 0255035 is a selective, competitive antagonist of the M1 muscarinic acetylcholine receptor used in preclinical research on central nervous system disorders. The compound (CAS 1135243-19-4) has molecular formula C18H20N6O3S2 and is supplied as a high-purity research-grade material for in vitro and in vivo pharmacology.
  • Selective M1 receptor antagonism for pharmacological and mechanistic studies.
  • High chemical purity suitable for analytical and biological assays.
  • Available in multiple small-quantity formats for flexibility in dosing and screening.
  • Suitable for both in vitro and in vivo experimental applications.
  • Stable when stored under recommended conditions for research use.
Catalog No. 50-002-42004 Supplier Medchemexpress LLC Supplier No. HY10823410MG
VU 0255035 is a selective, competitive antagonist of the M1 muscarinic acetylcholine receptor used in preclinical research on central nervous system disorders. The compound (CAS 1135243-19-4) has molecular formula C18H20N6O3S2 and is supplied as a high-purity research-grade material for in vitro and in vivo pharmacology.
  • Selective M1 receptor antagonism for pharmacological and mechanistic studies.
  • High chemical purity suitable for analytical and biological assays.
  • Available in multiple small-quantity formats for flexibility in dosing and screening.
  • Suitable for both in vitro and in vivo experimental applications.
  • Stable when stored under recommended conditions for research use.