Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[3-oxo-3-(4-pyridin-4-ylpiperazin-1-yl)propyl]-2,1,3-benzothiadiazole-4-sulfonamide | 1135243-19-4 | MFCD18086889 | 99.8% | 432.52 | C18H20N6O3S2 | 100 MG
SDP

Numéro de catalogue. 5000241984
Encompass_Preferred

VU 0255035 is a selective and competitive M1 muscarinic acetylcholine receptor (M1 mAChR) antagonist used as a research tool to probe M1 signaling and neuronal excitability in central nervous system disease models.

  • Selective, competitive M1 muscarinic receptor antagonist.
  • Brain-penetrant compound suitable for CNS studies.
  • High purity, approximately 99.8%.
  • Molecular weight 432.52 g/mol; chemical formula C18H20N6O3S2.
  • Appearance: solid, light yellow to brown; supplied as 100 MG quantities.
  • Used in research related to epilepsy, Parkinson's disease, and dystonia.

Numéro de catalogue. 50-002-41984 Fournisseur Medchemexpress LLC Code fournisseur HY108234100MG
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

VU 0255035 is a selective and competitive M1 muscarinic acetylcholine receptor (M1 mAChR) antagonist used as a research tool to probe M1 signaling and neuronal excitability in central nervous system disease models.

  • Selective, competitive M1 muscarinic receptor antagonist.
  • Brain-penetrant compound suitable for CNS studies.
  • High purity, approximately 99.8%.
  • Molecular weight 432.52 g/mol; chemical formula C18H20N6O3S2.
  • Appearance: solid, light yellow to brown; supplied as 100 MG quantities.
  • Used in research related to epilepsy, Parkinson's disease, and dystonia.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.