Learn More
Medchemexpress LLC N-[3-oxo-3-(4-pyridin-4-ylpiperazin-1-yl)propyl]-2,1,3-benzothiadiazole-4-sulfonamide | 1135243-19-4 | MFCD18086889 | 99.8% | 432.52 g·mol⁻¹ | C18H20N6O3S2 | 50 MG
SDP
Catalog No. 5000241985
Encompass_Preferred
VU 0255035 is a selective, brain-penetrant muscarinic M1 receptor antagonist used as a research tool in neuroscience and pharmacology. It acts as a competitive M1 antagonist with demonstrated in vitro and in vivo activity, including reduction of cholinergic-induced neuronal depolarization and efficacy in seizure models. The compound is supplied as a well-characterized, high-purity small molecule for laboratory use.
  • Selective muscarinic M1 receptor antagonist
  • Brain-penetrant small molecule suitable for in vivo studies
  • Demonstrated activity in electrophysiology and seizure models
  • High purity suitable for pharmacology experiments
  • Available in multiple laboratory-friendly package sizes
Catalog No. 50-002-41985 Supplier Medchemexpress LLC Supplier No. HY10823450MG
VU 0255035 is a selective, brain-penetrant muscarinic M1 receptor antagonist used as a research tool in neuroscience and pharmacology. It acts as a competitive M1 antagonist with demonstrated in vitro and in vivo activity, including reduction of cholinergic-induced neuronal depolarization and efficacy in seizure models. The compound is supplied as a well-characterized, high-purity small molecule for laboratory use.
  • Selective muscarinic M1 receptor antagonist
  • Brain-penetrant small molecule suitable for in vivo studies
  • Demonstrated activity in electrophysiology and seizure models
  • High purity suitable for pharmacology experiments
  • Available in multiple laboratory-friendly package sizes