Learn More
AdipoGen N-(3-Oxohexanoyl)-L-homoserine lactone
SDP
Catalog No. 501016163
Encompass_Preferred
N-(3-Oxohexanoyl)-L-homoserine lactone, AdipoGen Life Sciences. Chemical. CAS: 143537-62-6. Formula: C10H15NO4. MW: 213.2. Synthetic. N-(3-Oxohexanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. At increased populations of bacteria, localized higher concentrations of 3OC6-HSL, an endogenous ligand to transcriptional factor LuxR, leads to increased production of both the AHL synthase and proteins responsible for bioluminescence. Numerous other species of bacteria also employ 3OC6-HSL in cell-to-cell communication.
Catalog No. 50-101-6163 Supplier AdipoGen Supplier No. CDXO0057M010
Add to Cart
Add to Cart
N-(3-Oxohexanoyl)-L-homoserine lactone, AdipoGen Life Sciences. Chemical. CAS: 143537-62-6. Formula: C10H15NO4. MW: 213.2. Synthetic. N-(3-Oxohexanoyl)-L-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms. At increased populations of bacteria, localized higher concentrations of 3OC6-HSL, an endogenous ligand to transcriptional factor LuxR, leads to increased production of both the AHL synthase and proteins responsible for bioluminescence. Numerous other species of bacteria also employ 3OC6-HSL in cell-to-cell communication.