Learn More
Medchemexpress LLC N-(3-Phenylpropionyl)glycine-d2 | 1219795-43-3 | 5 MG
SDP
Catalog No. 5000417619
Encompass_Preferred
N-(3-Phenylpropionyl)glycine-d2 is the deuterium labeled N-(3-Phenylpropionyl)glycine. It can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitation during drug development.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-17619 Supplier Medchemexpress LLC Supplier No. HYN7403S5MG
N-(3-Phenylpropionyl)glycine-d2 is the deuterium labeled N-(3-Phenylpropionyl)glycine. It can be used as a tracer and an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer for quantitation during drug development.
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.