Learn More
Medchemexpress LLC N-(3-propylcarbamoyloxirane-2-carbonyl)-isoleucyl-proline | 134448-10-5 | MFCD00797531 | >98.0% | 383.44 g/mol | C18H29N3O6 | 100 MG
SDP
Catalog No. 5000244727
Encompass_Preferred
CA-074 is a potent, selective inhibitor of cathepsin B with a reported Ki of 2-5 nM, used in biochemical and cell-based research to study cysteine protease activity. It is an epoxysuccinyl peptide derivative with typical purity over 98% and a molecular weight of 383.44 g/mol.
  • Potent cathepsin B inhibition with Ki of 2-5 nM.
  • Suitable for biochemical and cell-based assays.
  • Typical purity greater than 98% (HPLC).
  • Molecular formula C18H29N3O6 and molecular weight 383.44 g/mol.
  • Available in multiple pack sizes, including 100 MG, for research use.
Catalog No. 50-002-44727 Supplier Medchemexpress LLC Supplier No. HY103350100MG
CA-074 is a potent, selective inhibitor of cathepsin B with a reported Ki of 2-5 nM, used in biochemical and cell-based research to study cysteine protease activity. It is an epoxysuccinyl peptide derivative with typical purity over 98% and a molecular weight of 383.44 g/mol.
  • Potent cathepsin B inhibition with Ki of 2-5 nM.
  • Suitable for biochemical and cell-based assays.
  • Typical purity greater than 98% (HPLC).
  • Molecular formula C18H29N3O6 and molecular weight 383.44 g/mol.
  • Available in multiple pack sizes, including 100 MG, for research use.