Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-[3-(Trimethoxysilyl)propyl]aniline

Catalog No. 44080950ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-080-950ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-080-950ML Supplier Sigma Aldrich Supplier No. 44080950ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-[3-(Trimethoxysilyl)propyl]aniline (TMSPA) is a silane based propylaniline that can be used as a conducting polymer (CP). It can modify the surface characteristics and exhibits hydrophobic properties. It also enhances the wettability, dispersion of nanomaterials and selective absorption.TMSPA is used as a surface modifying agent that enhances the wettability, dispersion of silica which enhances the anti-degradation in natural rubber (NR). It can be used to functionalize gold nanorods (GNRs) through enzymatic polymerization which may be used for electrochemical detection of hydrogen peroxide (H2O2). It can also be used as an amine based solvent that forms a stable dispersion of single walled carbon nanotubes(SWCNT) after centrifugation.

Specifications

CAS 3068-76-6
Density 1.07 g/mL (at 25°C)
Boiling Point 310°C
Molecular Formula C12H21NO3Si
Refractive Index n20/D 1.506 (literature)
Linear Formula C6H5NH(CH2)3 Si(OCH3)3
MDL Number MFCD00053673
Quantity 50 mL
Synonym Trimethoxy[3-(phenylamino)propyl]silane; (3-Anilinopropyl)trimethoxysilane; N-[3-(Trimethoxysilyl)propyl]aniline; [3-(Phenylamino)propyl]trimethoxysilane
Molecular Weight (g/mol) 255.39
Biological Activity Blood
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.