Learn More
Cayman Chemical N-(3-hydroxy-7-cis tetradecenoyl)-L-Homoserine lactone, Purity: ≥95%, CAS: 273734-65-9, 10 mg
SDP
Catalog No. 502893022
Encompass_Preferred
An AHL involved in quorum sensing; did not affect cytokine or antibody production in mouse and human leukocyte immunoassays using LPS-stimulated macrophages
Catalog No. 50-289-3022 Supplier Cayman Chemical Supplier No. 900293910 MG
An AHL involved in quorum sensing; did not affect cytokine or antibody production in mouse and human leukocyte immunoassays using LPS-stimulated macrophages