Learn More
Cayman Chemical N-(3-hydroxyphenyl)-Arachidonoyl amide, Purity: ≥98%, CAS: 183718-75-4, 5 mg
SDP
Catalog No. 502890125
Encompass_Preferred
An irreversible COX-2 inhibitor (IC50 = 2 µM)
Catalog No. 50-289-0125 Supplier Cayman Chemical Supplier No. 100077045 MG
An irreversible COX-2 inhibitor (IC50 = 2 µM)