Learn More
Cayman Chemical N-(3-hydroxyphenyl)-Arachidonoyl amide, Purity: ≥98%, CAS: 183718-75-4, 100 mg
SDP
Catalog No. 502888236
Encompass_Preferred
An irreversible COX-2 inhibitor (IC50 = 2 µM)
Catalog No. 50-288-8236 Supplier Cayman Chemical Supplier No. 10007704100 MG
An irreversible COX-2 inhibitor (IC50 = 2 µM)