Learn More
Cayman Chemical N 3PhenylpropIonylglycIn 100mg
SDP
Catalog No. 502948648
Encompass_Preferred
An acylglycine; urinary levels are increased in patients with MCAD deficiency
Catalog No. 50-294-8648 Supplier Cayman Chemical Supplier No. 34392100 MG
An acylglycine; urinary levels are increased in patients with MCAD deficiency