Learn More
Cayman Chemical N-(3-Phenylpropionyl)glycine, Purity: ≥98%, CAS: 56613-60-6, 500 mg
SDP
Catalog No. 502948620
Encompass_Preferred
An acylglycine; urinary levels are increased in patients with MCAD deficiency
Catalog No. 50-294-8620 Supplier Cayman Chemical Supplier No. 34392500 MG
An acylglycine; urinary levels are increased in patients with MCAD deficiency