Learn More
Medchemexpress LLC N-[(3R)-1-azabicyclo[2.2.2]octan-3-yl]furo[2,3-c]pyridine-5-carboxamide | 478149-53-0 | 100.0% | 271.32 | C15H17N3O2 | 5 MG
SDP
Catalog No. 5000240500
Encompass_Preferred
PHA-543613 is a selective, brain-penetrant small-molecule agonist of the α7 nicotinic acetylcholine receptor used as a research tool in neuroscience and pharmacology. Supplied as the free base in a white to off-white solid form, it is provided at high purity for in vitro and in vivo experiments.
  • Selective α7 nicotinic acetylcholine receptor agonist.
  • Brain-penetrant small molecule suitable for CNS studies.
  • High purity (reported ~99.95%) for reliable experimental results.
  • Available in small laboratory pack sizes for dose-ranging studies.
  • Supplied as a free base solid for straightforward handling and formulation.
Catalog No. 50-002-40500 Supplier Medchemexpress LLC Supplier No. HY1056705MG
PHA-543613 is a selective, brain-penetrant small-molecule agonist of the α7 nicotinic acetylcholine receptor used as a research tool in neuroscience and pharmacology. Supplied as the free base in a white to off-white solid form, it is provided at high purity for in vitro and in vivo experiments.
  • Selective α7 nicotinic acetylcholine receptor agonist.
  • Brain-penetrant small molecule suitable for CNS studies.
  • High purity (reported ~99.95%) for reliable experimental results.
  • Available in small laboratory pack sizes for dose-ranging studies.
  • Supplied as a free base solid for straightforward handling and formulation.