Learn More
Medchemexpress LLC N-[(3R,4S)-3-hydroxy-2,2-dimethyl-6-(4,4,4-trifluorobutoxy)chroman-4-yl]-N-methylmethanesulfonamide | 223749-46-0 | MFCD12756323 | 100.0% | 411.44 | C17H24F3NO5S | 10 MG
SDP
Catalog No. 5000244141
Encompass_Preferred
HMR 1556 is a chromanol-derived small molecule that selectively inhibits the I_Ks (slow delayed rectifier potassium) current. It exhibits potent activity in cardiac myocytes with reported IC50 values in the low nanomolar range and is supplied as a high-purity research reagent in solid and solution formats.
  • Potent and selective Iks channel blocker.
  • Reported IC50s of 10.5 nM (canine) and 34 nM (guinea pig).
  • High reported purity (99.95%).
  • Available as solid and as DMSO solution formats.
  • Multiple package sizes to suit experimental scale.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-44141 Supplier Medchemexpress LLC Supplier No. HY10636910MG
HMR 1556 is a chromanol-derived small molecule that selectively inhibits the I_Ks (slow delayed rectifier potassium) current. It exhibits potent activity in cardiac myocytes with reported IC50 values in the low nanomolar range and is supplied as a high-purity research reagent in solid and solution formats.
  • Potent and selective Iks channel blocker.
  • Reported IC50s of 10.5 nM (canine) and 34 nM (guinea pig).
  • High reported purity (99.95%).
  • Available as solid and as DMSO solution formats.
  • Multiple package sizes to suit experimental scale.
  • For research use only; not for human or clinical use.