Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(3S,4R)-6-cyano-3,4-dihydro-3-hydroxy-2,2-dimethyl-2H-1-benzopyran-4-yl]-N-hydroxy-acetamide | 127408-31-5 | 99.7% | 276.29 g/mol | C14H16N2O4 | 1 MG
SDP

Catalog No. 5000212065
Encompass_Preferred

Y-26763 is a small-molecule ATP-sensitive potassium (KATP) channel opener used in research. It is the active metabolite of Y-27152 and is supplied as a high-purity compound for in vitro and in vivo studies that investigate KATP channel modulation.

  • ATP-sensitive potassium (KATP) channel activator for electrophysiology and pharmacology studies.
  • High purity (99.7%) suitable for analytical and biological assays.
  • Molecular weight 276.29 g/mol and formula C14H16N2O4 for accurate dosing.
  • Documented CAS number (127408-31-5) for unambiguous identification.
  • Soluble in 1 M NaOH (27 mg/mL) with sonication/heat-assisted dissolution.
  • Available in milligram quantities for pilot experiments and screening.

Catalog No. 50-002-12065 Supplier Medchemexpress LLC Supplier No. HY1010691MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Y-26763 is a small-molecule ATP-sensitive potassium (KATP) channel opener used in research. It is the active metabolite of Y-27152 and is supplied as a high-purity compound for in vitro and in vivo studies that investigate KATP channel modulation.

  • ATP-sensitive potassium (KATP) channel activator for electrophysiology and pharmacology studies.
  • High purity (99.7%) suitable for analytical and biological assays.
  • Molecular weight 276.29 g/mol and formula C14H16N2O4 for accurate dosing.
  • Documented CAS number (127408-31-5) for unambiguous identification.
  • Soluble in 1 M NaOH (27 mg/mL) with sonication/heat-assisted dissolution.
  • Available in milligram quantities for pilot experiments and screening.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.