Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[[3S-[(propylamino)carbonyl]-2S-oxiranyl]carbonyl]-L-isoleucyl-L-proline | 134448-10-5 | MFCD00797531 | >98.0% | 383.44 g/mol | C18H29N3O6 | 50 MG
SDP

Catalog No. 5000244730
Encompass_Preferred

CA-074 is a potent, selective inhibitor of cathepsin B with a reported Ki of 2-5 nM (CAS 134448-10-5). It is supplied as a research-grade solid for biochemical and cell-based studies of cathepsin B-dependent processes.

  • Potent inhibitor of cathepsin B (Ki = 2-5 nM).
  • High purity by HPLC (>98%).
  • Molecular weight 383.44 g/mol.
  • Chemical formula C18H29N3O6.
  • White to off-white powder appearance.
  • Intended for research use only.

Catalog No. 50-002-44730 Supplier Medchemexpress LLC Supplier No. HY10335050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CA-074 is a potent, selective inhibitor of cathepsin B with a reported Ki of 2-5 nM (CAS 134448-10-5). It is supplied as a research-grade solid for biochemical and cell-based studies of cathepsin B-dependent processes.

  • Potent inhibitor of cathepsin B (Ki = 2-5 nM).
  • High purity by HPLC (>98%).
  • Molecular weight 383.44 g/mol.
  • Chemical formula C18H29N3O6.
  • White to off-white powder appearance.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.