Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-(1H-benzimidazol-2-yl)phenyl]-2,4-dimethoxybenzamide | 476633-98-4 | 99.7% | 373.40 g·mol⁻1 | C22H19N3O3 | 10 MG
SDP

Catalog No. 5000287716
Encompass_Preferred

Sirtuin modulator 6 is a small-molecule research compound that modulates sirtuin activity and is used in studies of lifespan, metabolic disorders, and neurodegenerative disease. The material is provided as a characterized solid with supporting batch documentation for research use in biochemical and cellular assays.

  • High HPLC purity (99.7%).
  • Molecular formula C22H19N3O3; molecular weight 373.40 g·mol⁻1.
  • CAS registry number 476633-98-4 confirms chemical identity.
  • Supplied in small research pack sizes suitable for assay preparation.
  • Characterized by batch-specific COA and SDS for safety and quality data.
  • Suitable for biochemical and cellular sirtuin pathway studies.

Catalog No. 50-002-87716 Supplier Medchemexpress LLC Supplier No. HY15598810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sirtuin modulator 6 is a small-molecule research compound that modulates sirtuin activity and is used in studies of lifespan, metabolic disorders, and neurodegenerative disease. The material is provided as a characterized solid with supporting batch documentation for research use in biochemical and cellular assays.

  • High HPLC purity (99.7%).
  • Molecular formula C22H19N3O3; molecular weight 373.40 g·mol⁻1.
  • CAS registry number 476633-98-4 confirms chemical identity.
  • Supplied in small research pack sizes suitable for assay preparation.
  • Characterized by batch-specific COA and SDS for safety and quality data.
  • Suitable for biochemical and cellular sirtuin pathway studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.