Learn More
Medchemexpress LLC N-[4-[(1S,2R)-2-aminocyclopropyl]phenyl]-4-(4-methylpiperazin-1-yl)benzamide dihydrochloride | 1831167-98-6 | 99.3% | 423.38 g·mol⁻¹ | C21H28Cl2N4O | 10 MG
SDP
Catalog No. 5000294766
Encompass_Preferred
DDP-38003 dihydrochloride is a small-molecule, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with a reported IC50 of 84 nM. Supplied as the dihydrochloride salt, the compound is provided with a characterized molecular formula and high purity for research use.
  • Orally available inhibitor of KDM1A/LSD1 with IC50 ≈84 nM.
  • Supplied as a dihydrochloride salt to improve handling and solubility.
  • High purity suitable for in vitro and in vivo research applications.
  • Well-characterized molecular weight and formula for experimental planning.
  • Available in multiple small-scale sizes to support laboratory workflows.
Catalog No. 50-002-94766 Supplier Medchemexpress LLC Supplier No. HY19612A10MG
DDP-38003 dihydrochloride is a small-molecule, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with a reported IC50 of 84 nM. Supplied as the dihydrochloride salt, the compound is provided with a characterized molecular formula and high purity for research use.
  • Orally available inhibitor of KDM1A/LSD1 with IC50 ≈84 nM.
  • Supplied as a dihydrochloride salt to improve handling and solubility.
  • High purity suitable for in vitro and in vivo research applications.
  • Well-characterized molecular weight and formula for experimental planning.
  • Available in multiple small-scale sizes to support laboratory workflows.