Learn More
Medchemexpress LLC N-[4-(2-amino-3-chloropyridin-4-yl)oxy-3-fluorophenyl]-5-(4-fluorophenyl)-4-oxo-1H-pyridine-3-carboxamide | 1174046-72-0 | MFCD18206786 | 99.5% | 468.84 g·mol-1 | C23H15ClF2N4O3 | 1 ML
SDP
Catalog No. 5000244444
Encompass_Preferred
BMS-794833 is a small-molecule kinase inhibitor active against VEGFR2 and Met, originally described in patent WO2009094417. It exhibits potent biochemical activity (VEGFR2 IC50 ≈ 15 nM; Met IC50 ≈ 1.7 nM) and is provided for research use in both solution and solid formats. Typical uses include biochemical assays, cell-based kinase studies, and preclinical pharmacology research.
  • Potent dual inhibition of VEGFR2 and Met with low-nanomolar IC50s.
  • High purity suitable for research applications.
  • Available as concentrated solution and solid quantities for flexibility.
  • Soluble in common organic solvents such as DMSO; verify before use.
  • Applicable to biochemical and cellular kinase assays and preclinical studies.
Catalog No. 50-002-44444 Supplier Medchemexpress LLC Supplier No. HY1049710MM 1ML
BMS-794833 is a small-molecule kinase inhibitor active against VEGFR2 and Met, originally described in patent WO2009094417. It exhibits potent biochemical activity (VEGFR2 IC50 ≈ 15 nM; Met IC50 ≈ 1.7 nM) and is provided for research use in both solution and solid formats. Typical uses include biochemical assays, cell-based kinase studies, and preclinical pharmacology research.
  • Potent dual inhibition of VEGFR2 and Met with low-nanomolar IC50s.
  • High purity suitable for research applications.
  • Available as concentrated solution and solid quantities for flexibility.
  • Soluble in common organic solvents such as DMSO; verify before use.
  • Applicable to biochemical and cellular kinase assays and preclinical studies.