Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-[(2-amino-6-methyl-4-pyrimidinyl)amino]phenyl]-4-(4-quinolinylamino)benzamide | 1020149-73-8 | MFCD27937047 | 99.8% | 461.52 g/mol | C27H23N7O | 5 MG
SDP

Catalog No. 5000222712
Encompass_Preferred

SGI-1027 is a non-nucleoside DNA methyltransferase (DNMT) inhibitor for research use that inhibits DNMT1, DNMT3A, and DNMT3B; reported IC50 values include 12.5 μM (DNMT1), 8 μM (DNMT3A), and 7.5 μM (DNMT3B).

  • Non-nucleoside DNMT inhibitor active against DNMT1, DNMT3A, and DNMT3B.
  • Reported IC50 values: DNMT1 12.5 μM, DNMT3A 8 μM, DNMT3B 7.5 μM.
  • High supplied purity (~99.8%).
  • Molecular formula C27H23N7O and molecular weight 461.52 g/mol.
  • Available as solid and as a 10 mM DMSO solution for convenient use.
  • Storage guidance provided for powder and solvent formats to preserve stability.

Catalog No. 50-002-22712 Supplier Medchemexpress LLC Supplier No. HY139625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SGI-1027 is a non-nucleoside DNA methyltransferase (DNMT) inhibitor for research use that inhibits DNMT1, DNMT3A, and DNMT3B; reported IC50 values include 12.5 μM (DNMT1), 8 μM (DNMT3A), and 7.5 μM (DNMT3B).

  • Non-nucleoside DNMT inhibitor active against DNMT1, DNMT3A, and DNMT3B.
  • Reported IC50 values: DNMT1 12.5 μM, DNMT3A 8 μM, DNMT3B 7.5 μM.
  • High supplied purity (~99.8%).
  • Molecular formula C27H23N7O and molecular weight 461.52 g/mol.
  • Available as solid and as a 10 mM DMSO solution for convenient use.
  • Storage guidance provided for powder and solvent formats to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.