Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-(2-chloro-5-methylpyrimidin-4-yl)phenyl]-N-[(4-(difluoroacetylamino)methyl)phenyl]methyl-2,4-dihydroxybenzamide | 1684386-71-7 | 99.1% | 552.96 g/mol | C28H23ClF2N4O4 | 25MG
SDP

Catalog No. 5000235093
Encompass_Preferred

VER-246608 is a potent, ATP-competitive inhibitor of pyruvate dehydrogenase kinase (PDK) isoforms, developed for biochemical and cell-based research. It shows sub-100 nM enzymatic potency across PDK isoforms and reduces E1α Ser293 phosphorylation in cellular assays. Supplied as a solid and as DMSO solutions for laboratory use.

  • Potent ATP-competitive inhibition of pyruvate dehydrogenase kinase isoforms.
  • IC50 values: PDK-1 35 nM; PDK-3 40 nM; PDK-2 84 nM; PDK-4 91 nM.
  • Suppresses phosphorylation of E1α Ser293 in cellular assays (IC50 ≈ 266 nM).
  • High purity: 99.1%.
  • Molecular formula C28H23ClF2N4O4; molecular weight 552.96 g/mol.
  • Available as solid (25 MG) and as 10 mM solutions in DMSO for convenience.
  • Recommended storage: powder at -20°C or 4°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-35093 Supplier Medchemexpress LLC Supplier No. HY1249225MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

VER-246608 is a potent, ATP-competitive inhibitor of pyruvate dehydrogenase kinase (PDK) isoforms, developed for biochemical and cell-based research. It shows sub-100 nM enzymatic potency across PDK isoforms and reduces E1α Ser293 phosphorylation in cellular assays. Supplied as a solid and as DMSO solutions for laboratory use.

  • Potent ATP-competitive inhibition of pyruvate dehydrogenase kinase isoforms.
  • IC50 values: PDK-1 35 nM; PDK-3 40 nM; PDK-2 84 nM; PDK-4 91 nM.
  • Suppresses phosphorylation of E1α Ser293 in cellular assays (IC50 ≈ 266 nM).
  • High purity: 99.1%.
  • Molecular formula C28H23ClF2N4O4; molecular weight 552.96 g/mol.
  • Available as solid (25 MG) and as 10 mM solutions in DMSO for convenience.
  • Recommended storage: powder at -20°C or 4°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.