Learn More
Medchemexpress LLC N-{4-[(2-chloro-6-fluorobenzyl)oxy]-3-methoxybenzyl}-2-(morpholin-4-yl)ethanamine | 875005-43-9 | 99.6% | C21H26ClFN2O3 | 10MG
SDP
Catalog No. 5000259354
Encompass_Preferred
GLP-1R modulator C16 is a small-molecule allosteric modulator that enhances glucagon-like peptide-1 (GLP-1) binding to the GLP-1 receptor via a transmembrane site. Supplied for research use in solid form or as a DMSO solution, it is characterized by micromolar potency and high reported purity, and is intended for in vitro pharmacology and receptor modulation studies.
  • Enhances GLP-1 binding to the GLP-1 receptor via a transmembrane allosteric site.
  • Micromolar potency with EC50 ≈ 8.43 ± 3.82 μM.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C (long term).
Catalog No. 50-002-59354 Supplier Medchemexpress LLC Supplier No. HY14183910MG
GLP-1R modulator C16 is a small-molecule allosteric modulator that enhances glucagon-like peptide-1 (GLP-1) binding to the GLP-1 receptor via a transmembrane site. Supplied for research use in solid form or as a DMSO solution, it is characterized by micromolar potency and high reported purity, and is intended for in vitro pharmacology and receptor modulation studies.
  • Enhances GLP-1 binding to the GLP-1 receptor via a transmembrane allosteric site.
  • Micromolar potency with EC50 ≈ 8.43 ± 3.82 μM.
  • High purity suitable for biochemical and cell-based assays.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Recommended storage: powder -20°C (long term) or 4°C; in solvent -80°C (long term).