Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[4-[2-ethyl-4-(3-methylphenyl)-5-thiazolyl]-2-pyridinyl]benzamide | 303162-79-0 | MFCD17012805 | 99.8% | 399.51 g/mol | C24H21N3OS | 200 MG
SDP

Catalog No. 5000243212
Encompass_Preferred

TAK-715 is an orally active, potent p38 MAPK inhibitor used in research to probe p38α signaling and related pathways; it also inhibits casein kinase I (CK1δ/ε) and has demonstrated efficacy in preclinical inflammation models.

  • Potent p38α inhibition (IC50 7.1 nM) with reduced activity against p38β (IC50 200 nM).
  • Reported inhibition of CK1δ/ε and modulation of Wnt/β-catenin signaling.
  • Chemical formula C24H21N3OS; molecular weight 399.51 g/mol.
  • High reported purity (99.76%).
  • Soluble in DMSO (≈50 mg/mL) with suggested protocols for in vivo formulation.
  • Powder stable at -20°C for extended storage; solvent stocks stable at -80°C.

Catalog No. 50-002-43212 Supplier Medchemexpress LLC Supplier No. HY10456200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TAK-715 is an orally active, potent p38 MAPK inhibitor used in research to probe p38α signaling and related pathways; it also inhibits casein kinase I (CK1δ/ε) and has demonstrated efficacy in preclinical inflammation models.

  • Potent p38α inhibition (IC50 7.1 nM) with reduced activity against p38β (IC50 200 nM).
  • Reported inhibition of CK1δ/ε and modulation of Wnt/β-catenin signaling.
  • Chemical formula C24H21N3OS; molecular weight 399.51 g/mol.
  • High reported purity (99.76%).
  • Soluble in DMSO (≈50 mg/mL) with suggested protocols for in vivo formulation.
  • Powder stable at -20°C for extended storage; solvent stocks stable at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.